Skip to Content
TEM-0300-CU
Veco Speciment Grid
Copper, size 300 Square mesh , with Center Reference, vial of 100
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41111761
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
vial of 100
size
300 Square mesh , with Center Reference
General description
Square Mesh with Center Reference. Copper Grids, 3.05 mm diameter, 0.8 mil thickness. Pitch 83, hole 45, bar 38 micron, 300 mesh Cu. Veco Grids are the most rigid grids available.
Choose from one of the most recent versions:
Certificates of Analysis (COA)