MilliporeSigma
All Photos(1)

W256900

Sigma-Aldrich

α-Hexylcinnamaldehyde

≥95%, stabilized, FG

Synonym(s):
Hexyl cinnamic aldehyde
Linear Formula:
C6H5CH=C[(CH2)5CH3]CHO
CAS Number:
Molecular Weight:
216.32
FEMA Number:
2569
Council of Europe no.:
129
MDL number:
PubChem Substance ID:
Flavis number:
5.041
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

assay

≥95%

contains

0.075-0.500% BHT as stabilizer

composition

Contains IFRA and EU1223/2009 α-Hexylcinnamaldehyde

refractive index

n20/D 1.55 (lit.)

bp

174-176 °C/15 mmHg (lit.)

density

0.95 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

α-Hexylcinnamaldehyde

Organoleptic

green; floral; sweet

SMILES string

[H]C(=O)\C(CCCCCC)=C\c1ccccc1

InChI

1S/C15H20O/c1-2-3-4-6-11-15(13-16)12-14-9-7-5-8-10-14/h5,7-10,12-13H,2-4,6,11H2,1H3/b15-12+

InChI key

GUUHFMWKWLOQMM-NTCAYCPXSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

α-Hexylcinnamaldehyde is commonly used as a flavoring and fragrance ingredient. It shows potential chemosensitizing properties.

Packaging

1 kg in poly bottle
5, 10 kg in poly drum

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service