W259519

Sigma-Aldrich

β-Ionone

natural, ≥85%, FG

Synonym(s):
4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one
Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
FEMA Number:
2595
Beilstein/REAXYS Number:
1909544
EC Number:
Council of Europe no.:
142
MDL number:
PubChem Substance ID:
Flavis number:
7.008

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

assay

≥85%

refractive index

n20/D 1.52 (lit.)

bp

126-128 °C/12 mmHg (lit.)

density

0.945 g/mL at 25 °C (lit.)

Featured Industry

Flavors and Fragrances

SMILES string

CC(=O)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h7-8H,5-6,9H2,1-4H3/b8-7+

InChI key

PSQYTAPXSHCGMF-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural Occurrence: Tobacco, grape, grape brandy, orange juice, papaya, peach, raspberry, roasted almond, tea and spearmint.

Biochem/physiol Actions

Taste at 1-20 ppm

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3082 9 / PGIII

WGK Germany

2

Flash Point(F)

>235.4 °F

Flash Point(C)

>113 °C

Ming Ma et al.
Chembiochem : a European journal of chemical biology, 12(1), 88-99 (2010-12-15)
CYP101C1 from Novosphingobium aromaticivorans DSM12444 is a homologue of CYP101D1 and CYP101D2 enzymes from the same bacterium and CYP101A1 from Pseudomonas putida. CYP101C1 does not bind camphor but is capable of binding and hydroxylating ionone derivatives including α- and β-ionone...
Sivakumar Sekharan et al.
Journal of the American Chemical Society, 132(45), 15856-15859 (2010-10-23)
Visual pigment rhodopsin provides a decisive crossing point for interaction between organisms and environment. Naturally occurring visual pigments contain only PSB11 and 3,4-dehydro-PSB11 as chromophores. Therefore, the ability of visual opsin to discriminate between the retinal geometries is investigated by...
Mun-Ock Kim et al.
Molecular cancer therapeutics, 9(4), 833-843 (2010-04-01)
beta-Ionone (ION), an end-ring analogue of beta-carotenoid, has been known to inhibit tumor cell growth and induce apoptosis in various types of cancer cells. Nevertheless, its apoptosis-related molecular mechanisms remain unclear. Here, we first investigated the molecular mechanisms by which...
Jihai Shao et al.
Aquatic toxicology (Amsterdam, Netherlands), 104(1-2), 48-55 (2011-05-06)
In order to explore the potential targets of toxicity of β-ionone on the photosynthetic system of Microcystis aeruginosa, the polyphasic rise in chlorophyll a (Chl a) fluorescence transient and transcript expression for key genes in photosystem II (PSII) of M....
Vishal Sharma et al.
Bioorganic & medicinal chemistry letters, 22(20), 6343-6346 (2012-09-25)
A series of chalcones (3a-v) have been synthesized by condensation of β-ionone (1) with a variety of aldehydes (2a-v). The synthesized compounds have been screened for their in vitro antimicrobial activity against five bacterial and five fungal strains, using disc...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.