MilliporeSigma
All Photos(1)

W265616

Sigma-Aldrich

Maltol solution

1 wt. % in benzyl alcohol

Synonym(s):
3-Hydroxy-2-methyl-4-pyrone solution
Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
FEMA Number:
2656
MDL number:
PubChem Substance ID:
Flavis number:
7.014

biological source

synthetic

concentration

1 wt. % in benzyl alcohol

refractive index

n20/D 1.541

bp

205 °C

density

1.046 g/mL at 25 °C

application(s)

flavors and fragrances

Organoleptic

sweet

SMILES string

CC1=C(O)C(=O)C=CO1

InChI

1S/C6H6O3/c1-4-6(8)5(7)2-3-9-4/h2-3,8H,1H3

InChI key

XPCTZQVDEJYUGT-UHFFFAOYSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service