All Photos(1)

W265721

Sigma-Aldrich

Mandarin oil

Argentina

CAS Number:
FEMA Number:
2657
MDL number:

biological source

Citrus reticulata Blanco

optical activity

[α]20/D +89°, neat

refractive index

n20/D 1.474

bp

178 °C (lit.)

density

0.845 g/mL at 25 °C

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

Organoleptic

grape; orange; waxy; peach; sour

food allergen

no known allergens

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste @ 5 ppm

Certificate of Analysis

Certificate of Origin

Mosciano, G.
Perfumer & Flavorist, 28, 78-78 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service