W266309

Sigma-Aldrich

Marjoram oil

sweet, Certified organic (NOP)

CAS Number:
FEMA Number:
2663
MDL number:

Quality Level

biological source

Organum majorana

grade

Certified organic (NOP)
Halal
Kosher

reg. compliance

FDA 21 CFR 117

origin

Egypt origin

refractive index

n20/D 1.463

density

0.909 g/mL at 25 °C

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

Organoleptic

herbaceous; sweet

food allergen

no known allergens

Related Categories

Packaging

1 kg in glass bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Legal Information

Certified organic by MOSA

Pictograms

FlameHealth hazard

Signal Word

Danger

hazcat

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2

storage_class_code

3 - Flammable liquids

WGK Germany

WGK 3

Flash Point(F)

125.1 °F - closed cup

Flash Point(C)

51.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service