MilliporeSigma
All Photos(1)

W306802

Sigma-Aldrich

m-Tolualdehyde

Synonym(s):
3-Methylbenzaldehyde
Linear Formula:
CH3C6H4CHO
CAS Number:
Molecular Weight:
120.15
FEMA Number:
3068
Beilstein:
741964
EC Number:
MDL number:
PubChem Substance ID:
Flavis number:
5.028

biological source

synthetic

refractive index

n20/D 1.541 (lit.)

bp

199 °C (lit.)
80-82 °C/11 mmHg (lit.)

density

1.021 g/mL at 20 °C
1.019 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

SMILES string

[H]C(=O)c1cccc(C)c1

InChI

1S/C8H8O/c1-7-3-2-4-8(5-7)6-9/h2-6H,1H3

InChI key

OVWYEQOVUDKZNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service