W344001

Sigma-Aldrich

3-Nonanone

>96%, FG

Synonym(s):
Ethyl hexyl ketone
Linear Formula:
CH3(CH2)5COC2H5
CAS Number:
Molecular Weight:
142.24
FEMA Number:
3440
Beilstein/REAXYS Number:
1700447
EC Number:
Council of Europe no.:
11160
MDL number:
PubChem Substance ID:
Flavis number:
7.113
NACRES:
NA.21

Quality Level

biological source

Ricinus communis
Saccharum officinarum (sugarcane)

grade

FG
Halal
Kosher

Agency/Method

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

assay

>96%

refractive index

n20/D 1.42 (lit.)

bp

187-188 °C (lit.)

density

0.821 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

Organoleptic

cheese; green; fruity

food allergen

no known allergens

fragrance allergen

no known allergens

SMILES string

CCCCCCC(=O)CC

InChI

1S/C9H18O/c1-3-5-6-7-8-9(10)4-2/h3-8H2,1-2H3

InChI key

IYTXKIXETAELAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

25, 100 g in glass bottle
1 kg in glass bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
Peter S Toth et al.
The Analyst, 140(6), 1947-1954 (2015-02-11)
Two-phase voltammetry has been carried out using a reverse cell configuration, i.e. with the lower density organic solvent on the top of the aqueous solution in the cell, where the organic solvents contain either nitrile or ketone functional groups. The...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service