W350710

Sigma-Aldrich

Isoamyl isobutyrate

natural, 98%, FG

Linear Formula:
(CH3)2CHCH2CH2O2CCH(CH3)2
CAS Number:
Molecular Weight:
158.24
FEMA Number:
3507
EC Number:
MDL number:
PubChem Substance ID:
Flavis number:
9.419
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

assay

98%

refractive index

n20/D 1.406 (lit.)

bp

169-171 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

Featured Industry

Flavors and Fragrances

Organoleptic

green; waxy; fruity; sweet

food allergen

no known allergens

SMILES string

CC(C)CCOC(=O)C(C)C

InChI

1S/C9H18O2/c1-7(2)5-6-11-9(10)8(3)4/h7-8H,5-6H2,1-4H3

InChI key

VFTGLSWXJMRZNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

100 g in glass bottle
1 kg in glass bottle

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 2620 3 / PGIII

WGK Germany

WGK 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.