All Photos(1)

W516708

Sigma-Aldrich

Allyl isoamyl glycolate

≥98%

Empirical Formula (Hill Notation):
C10H18O3
CAS Number:
Molecular Weight:
186.25
EC Number:
MDL number:
PubChem Substance ID:

assay

≥98%

refractive index

n20/D 1.4317 (lit.)

bp

206-226 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCOCC(=O)OCC=C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service