MilliporeSigma
All Photos(1)

Z127132

Cochranes atom centers for Orbit molecular models

oxygen, carbonyl, red

Synonym(s):
chemistry model sets, molecular model sets

manufacturer/tradename

Cochranes 0218

color

red

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service