MilliporeSigma
All Photos(3)

Z155543

Sigma-Aldrich

Griffin-type SS beaker

capacity 1,200 mL, without handle

NACRES:
NB.43

material

stainless steel

feature

graduations: no

H × O.D.

154 mm × 105 mm

capacity

1,200 mL

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service