MilliporeSigma
All Photos(3)

Z155608

Griffin-type SS beaker

capacity 2,000 mL, with handle

NACRES:
NB.43

material

stainless steel

feature

graduations: no

H × O.D.

168 mm × 129 mm

capacity

2,000 mL

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service