Documents

Z185507

Cochranes atom centers for Orbit molecular models

iron, octahedral, dark gray

Synonym(s):
chemistry model sets, molecular model sets

manufacturer/tradename

Cochranes 0256

color

dark gray

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service