All Photos(1)
Z259810
Igloo® all-plastic cooler
capacity 3 gal (11.4 L)
H × I.D.
13 1/4 in. × 10 1/8 in.
capacity
3 gal (11.4 L)
General description
Industrial grade, HDPE yellow body, red lid, molded-in handles, Ultratherm® insulation, and recessed plastic spigots. Suitable for temporary storage of ice water, dry ice, or for use as hot/cold bath. White FDA grade liner resists stains and odors. 2 and 3 gal sizes have plastic bail handle. 5 and 10 gal coolers have plastic side handles and nylon cords to secure lid to side handle.
Legal Information
Igloo is a registered trademark of Igloo Products Corp.
Ultratherm is a registered trademark of Igloo Products Corp.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).