MilliporeSigma
All Photos(1)

Z311049

Mettler Toledo portable pH/ion meter

Model MP125, meter with Inlab® 413 IP67 waterproof electrode

grade

CE Compliant

description

BNC connector
electrode condition display
multipoint calibration: display
pH/mV/T functions

manufacturer/tradename

Model MP125

availability

available only in EU

mV measuring range

1,999

pH measuring range

−2.00-16.00, accuracy: 0.01, resolution: 0.01

parameter

−5-105 °C temperature

size

50 mm × 90 mm × 210 mm

battery power

AA

Looking for similar products? Visit Product Comparison Guide

General description

Meters ultrasonically welded for strength and waterproofing to IP67. Will float if dropped in water. Battery powered (batteries supplied) with approximately 500h battery life and auto shut-off after 10 min of non-use.

Features and Benefits

Excellent value with autostability and storage for 50 readings.

Legal Information

InLab is a registered trademark of Mettler-Toledo, Inc.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service