Documents
Z408999
MIXXOR liquid-liquid extraction system
capacity 50 mL
Pricing and availability is not currently available.
capacity
50 mL
General description
Sample Prep Benefits
  • For sample volumes from 2 to 50 mL
  • Minimal amount of solvent required
  • Rapid, simple-to-use
  • Safe, closed system prevents spills
  • Precise, allows for easy separation of phases
  • Flexible, comes in five sizes and will fit in interlocking stands
The MIXXOR Concept (seven easy steps)
1. Introduce sample and extraction solvent into reservoir A (Fig. 1)
2. Insert mixer-separator piston B into reservoir A, tighten cap S.
3. Pump four or more times to mix (Fig. 2)
4. Pull mixer-separator up slightly above liquid level and secure with holder spacer H. Loosen cap.
5. After separation, slide down the mixer-separator to transfer the upper phase into collection chamber E.
6. Adjust lower phase to top of axial channel C. (Fig. 1 and 3) Use fine adjustment on holder-spacer to secure setting.
7. Top phase can now be decanted safely, while lower phase stays in the axial chamber.
For the quantitative extraction of organics from aqueous solutions and separations in immunoassays. The MIXXOR system has been applied successfully in many laboratory solvent extraction operations and is ideal for the rapid screening of alternative solvents for specific extraction problems. System includes plastic support stand. See the website at sigma-aldrich.com/labware for a listing of replacement parts.
Legal Information
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.