Documents
Z544175
Molymod® Atoms and Bonds
Nitrogen, no. of holes, 4, diam. 23 mm, angle, 109.50
NACRES:
NB.17
feature
angle 109.50
no. of holes 4
manufacturer/tradename
Molymod
diam.
23 mm
color
blue
Packaging
1PAK = 10EA
10PAK = 10 x 1PAK or 100EA
Legal Information
Molymod is a registered trademark of Spiring Enterprises Ltd.
Certificate of Analysis
Certificate of Origin