All Photos(1)
Z632449
Retsch test sieves
40 μm, D × H 200 mm × 50 mm
material
stainless steel (with wire gauze)
availability
not available in USA and Canada
D × H
200 mm × 50 mm
particle size
40 μm
General description
For mechanical or manual, wet or dry sieving. Quintuple tested and provided with Quality Certificate DIN ISO 3310/1.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).