All Photos(1)

Z645079

IKA® KMO 2 basic IKAMAG stirrers

compact magnetic stirrer without heating element, stainless steel, 115V

Synonym(s):
magnetic stirrers, laboratory stirrers, lab stirrers
NACRES:
NB.73

AC/DC input

115 V AC, US 2-pin plug

material

stainless steel plate

feature

CSA approved

packaging

pack of 1

availability

not available in EU

parameter

0-1100 L/min speed
50-500 °C temp. range

overall W × H × D

140 mm × 75 mm × 200 mm

plate L × W

140 mm × 120 mm

Looking for similar products? Visit Product Comparison Guide

General description

IKA® KMO 2 Basic IKAMAG Stirrer

Small, powerful magnetic stirrer without heating element.

  • Strong magnetic field
  • Motor with optoelectronic speed control
  • Infinitely variable speed from 0-1100 rpm
  • Stainless steel casing facilitates cleaning and sterilization
  • Includes M 10 thread for H 16 V support rod

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG
IKAMAG is a trademark of IKA-Werke GmbH & Co. KG

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service