Documents

Z647462

Pyrex® measuring cylinder with spout, heavy duty, squat form

volume 250 mL, accuracy: 2.0 mL

NACRES:
NB.43
Pricing and availability is not currently available.

feature

graduations 5.0 mL

volume measuring range

250 mL, accuracy: 2.0 mL

H

212 mm

Looking for similar products? Visit Product Comparison Guide

General description

Heavy duty construction includes: thicker walls, heavily beaded rim and spout, and robust hexagonal foot.
  • Squat height is more stable
  • More convenient to use in confined spaces
  • Graduations in blue enamel

Legal Information

PYREX is a registered trademark of Corning, Inc.
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.