Documents
Z677248
Dermatril LS nitrile gloves
size XXL
material
blue gloves
blue nitrile
size
XXL
sterility
sterile
feature
CE compliant
latex free
powder-free: no
packaging
pack of 1 ea
manufacturer/tradename
KCL 747
L × thickness
280 mm × 0.11 mm
General description
Dermatril LS is a sterile, extended cuff, blue nitrile, lightly powdered glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril LS disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril LS disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
  • Individually packed in pairs of sterile gloves
  • Antistatic
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • Roughened fingertips for better grip
  • Rolled cuff edge for easier donning
  • CE compliant
  • Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 61340-5-1, ASTM 1671:2001
Legal Information
Dermatril is a trademark of KCL GmbH
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.