11814524001

Roche

rGFP

Green Fluorescent Protein from jellyfish Aequorea victoria, recombinant expressed in E. coli

Synonym(s):
green fluorescent protein, gfp, r-
Pricing and availability is not currently available.

Quality Level

form

solution

packaging

pkg of 50 μL (1 mg/ml)

mfr. no.

Roche

shipped in

wet ice

storage temp.

2-8°C

General description

GFP isolated from jellyfish Aequorea Victoria exhibited spontaneous fluorescence property. It is a novel important marker for gene expression in various heterologous systems. It is stably present in plant cells and shows photobleaching. Molecular cloning of GFP produces a fused protein with fluorescence excitation and emission spectra.

Specificity

Heat inactivation: Fluorescence will be quenched by reducing reagents, such as DTT or sodiumdithiosulfate, and is completely inhibited by peroxide.
Fluorescence is inhibited by extreme pH conditions (pH < 4 and pH > 11) as well as ethanol content higher than 60%.

Application

Recombinant green fluorescent protein (rGFP) has also been used in indirect enzyme-linked immunosorbent assay.
Recombinant Green Fluorescent Protein (rGFP) is suitable as:
  • Positive control when detecting GFP fusion proteins by SDS-PAGE.
  • Positive control when detecting GFP fusion proteins by western blot using Anti-GFP monoclonal antibody.
  • Standard for experiments involving fluorescence microscopy.

Physical form

Solution, 1 mg/ml rGFP in 5 mM Tris-HCl, 5 mM EDTA, pH 8.0

Preparation Note

Working concentration: Amount of rGFP recommended for:
  • SDS-PAGE (Coomassie stained): 1 μg
  • Western blot: 5 ng

Other Notes

Emission max: 509nm
Excitation max: 395nm
For life science research only. Not for use in diagnostic procedures.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Certificate of Analysis
Certificate of Origin
Efficient central nervous system AAVrh10-mediated intrathecal gene transfer in adult and neonate rats.
Hordeaux J, et al.
Gene Therapy, 22(4), 316-316 (2015)
S Inouye et al.
FEBS letters, 341(2-3), 277-280 (1994-03-21)
Expression of the cDNA for Aequorea green fluorescent protein in E. coli yielded a fused protein with fluorescence excitation and emission spectra virtually identical to those of the native green fluorescent protein. Further, a solution of the protein, when mixed...
J Sheen et al.
The Plant journal : for cell and molecular biology, 8(5), 777-784 (1995-11-01)
The green-fluorescent protein (GFP) from jellyfish Aequorea victoria has been used as a convenient new vital marker in various heterologous systems. However, it has been problematic to express GFP in higher eukaryotes, especially in higher plants. This paper reports that...
Methods for addressing the protein-protein interaction between histone deacetylase 6 and ubiquitin
dos Santos Passos C, et al.
bioRxiv, 203372-203372 (2017)
Articles
Biomaterials science involves the design and fabrication of smart materials for studying, directing, or mimicking biology. For successful integration of biomaterials in biological research, a meaningful understanding of biological systems is required.
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.