18-1580

Sigma-Aldrich

Lithium nitrate

SAJ special grade

Linear Formula:
LiNO3
CAS Number:
Molecular Weight:
68.95
EC Number:
MDL number:
PubChem Substance ID:

grade

SAJ special grade

form

crystalline

availability

available only in Japan

pH

7-9 (20 °C, 50 g/L)

mp

264 °C (lit.)

SMILES string

[Li+].[O-][N+]([O-])=O

InChI

1S/Li.NO3/c;2-1(3)4/q+1;-1

InChI key

IIPYXGDZVMZOAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

hazcat

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3

storage_class_code

5.1B - Oxidizing hazardous materials

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Sung-Ho Shin et al.
ACS nano, 8(10), 10844-10850 (2014-09-30)
We present a method to develop high performance flexible piezoelectric nanogenerators (NGs) by employing Li-doped ZnO nanowires (NWs). We synthesized Li-doped ZnO NWs and adopted them to replace intrinsic ZnO NWs with a relatively low piezoelectric coefficient. When we exploited...
Ghina Ali et al.
Biotechnology progress, 31(4), 883-889 (2015-04-29)
The immobilization of pullulanase from Klebsiella pneumoniae by grafting was investigated. Pullulanase was linked after activation of alginate via a covalent bond between the amine groups of the enzyme and the carboxylic acid groups of alginate. The immobilization yield was...
Xingwen Yu et al.
ACS applied materials & interfaces, 7(30), 16625-16631 (2015-07-15)
Lithium-sulfur (Li-S) batteries are receiving great attention as the most promising next-generation power source with significantly high charge-storage capacity. However, the implementation of Li-S batteries is hampered by a critical challenge because of the soluble nature of the intermediate polysulfide...
Soumyadip Choudhury et al.
ACS nano, 9(6), 6147-6157 (2015-05-28)
This study reports on a facile approach to the fabrication of nanoporous carbon cathodes for lithium sulfur batteries using gyroid carbon replicas based on use of polystyrene-poly-4-vinylpyridine (PS-P4VP) block copolymers as sacrificial templates. The free-standing gyroid carbon network with a...
Hoon Kim et al.
Nature communications, 6, 7278-7278 (2015-06-13)
Elemental sulfur is one of the most attractive cathode active materials in lithium batteries because of its high theoretical specific capacity. Despite the positive aspect, lithium-sulfur batteries have suffered from severe capacity fading and limited rate capability. Here we report...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service