MilliporeSigma
All Photos(1)

00044

Sigma-Aldrich

Boc-N-Me-Thr(Bzl)-OH

≥98.0% (HPLC)

Synonym(s):
Boc-O-benzyl-N-methyl-L-threonine
Empirical Formula (Hill Notation):
C17H25NO5
CAS Number:
Molecular Weight:
323.38
Beilstein:
6068811
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0% (HPLC)

form

powder or crystals

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

white

mp

65.4-71.4 °C

application(s)

peptide synthesis

SMILES string

C[C@@H](OCc1ccccc1)[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O

InChI

1S/C17H25NO5/c1-12(22-11-13-9-7-6-8-10-13)14(15(19)20)18(5)16(21)23-17(2,3)4/h6-10,12,14H,11H2,1-5H3,(H,19,20)/t12-,14+/m1/s1

InChI key

DMYNYZJGZSJUGU-OCCSQVGLSA-N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service