MilliporeSigma
All Photos(1)

03420590

α-Terpineol

primary reference standard

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Enzyme Commission number:
233-986-8
Beilstein:
2325137
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

storage temp.

−20°C

SMILES string

CC1=CC[C@H](CC1)C(C)(C)O

InChI

1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3

InChI key

WUOACPNHFRMFPN-UHFFFAOYSA-N

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service