MilliporeSigma
All Photos(1)

03535

Supelco

N-Pivaloylglycine

analytical standard

Synonym(s):
2-[(2,2-Dimethylpropanoyl)amino]acetic acid, N-(2,2,2-Trimethylacetyl)glycine, N-(2,2-Dimethyl-1-oxopropyl)glycin, Pivaloyl glycine
Empirical Formula (Hill Notation):
C7H13NO3
CAS Number:
Molecular Weight:
159.18
Beilstein:
1768401
MDL number:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

InChI

1S/C7H13NO3/c1-7(2,3)6(11)8-4-5(9)10/h4H2,1-3H3,(H,8,11)(H,9,10)/p-1

InChI key

RTRUEBYROMJFNU-UHFFFAOYSA-M

Biochem/physiol Actions

Pivaloyl glycine is an acylglycine. Acylglycines possess a common amidoacetic acid moiety and are normally minor metabolites of fatty acids. Elevated levels of certain acylglycines appear in the urine and blood of patients with various fatty acid oxidation disorders. They are normally produced through the action of glycine N-acyltransferase which is an enzyme that catalyzes the chemical reaction: acyl-CoA + glycine ↔ CoA + N-acylglycine.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service