MilliporeSigma
All Photos(1)

03667

Sigma-Aldrich

Boc-β-Homogln-OH

≥98.0% (TLC)

Synonym(s):
(S)-3-(Boc-amino)adipic acid 6-amide, Nβ-Boc-L-β-homoglutamine
Empirical Formula (Hill Notation):
C11H20N2O5
CAS Number:
Molecular Weight:
260.29
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

white

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCC(N)=O)CC(O)=O

InChI

1S/C11H20N2O5/c1-11(2,3)18-10(17)13-7(6-9(15)16)4-5-8(12)14/h7H,4-6H2,1-3H3,(H2,12,14)(H,13,17)(H,15,16)/t7-/m0/s1

InChI key

DAUHTFNYHSOVRQ-ZETCQYMHSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service