MilliporeSigma
All Photos(1)

04301

Supelco

Delphinidin 3-O-β-D-galactoside chloride

analytical standard

Synonym(s):
Delphinidin 3-O-β-D-galactopyranoside, Delphinidin 3-galactoside chloride, Empetrin chloride
Empirical Formula (Hill Notation):
C21H21ClO12
CAS Number:
Molecular Weight:
500.84
MDL number:
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC1=CC(O)=C(C=C(O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)C(C3=CC(O)=C(O)C(O)=C3)=[O+]4)C4=C1.[Cl-]

InChI

1S/C21H20O12.ClH/c22-6-15-17(28)18(29)19(30)21(33-15)32-14-5-9-10(24)3-8(23)4-13(9)31-20(14)7-1-11(25)16(27)12(26)2-7;/h1-5,15,17-19,21-22,28-30H,6H2,(H4-,23,24,25,26,27);1H/t15-,17+,18+,19-,21-;/m1./s1

InChI key

ZJWIIMLSNZOCBP-KGDMUXNNSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service