MilliporeSigma
All Photos(1)

04406

Supelco

Poly(styrenesulfonic acid sodium salt)

standard, 10,000, for GPC

Synonym(s):
Poly(sodium styrenesulfonate)
CAS Number:
MDL number:
NACRES:
NA.24

grade

analytical standard
for GPC
standard

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mp ~10200
Mw ~10600
Mw/Mn ≤1.20

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service