MilliporeSigma
All Photos(1)

05329

Sigma-Aldrich

(R)-4-Isopropylthiazolidine-2-thione

≥98.0% (GC)

Empirical Formula (Hill Notation):
C6H11NS2
CAS Number:
Molecular Weight:
161.29
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0% (GC)

optical activity

[α]20/D +37±3°, c = 1 in chloroform

optical purity

ee: ≥99:1 (LC)

mp

69-71 °C

SMILES string

CC(C)[C@@H]1CSC(=S)N1

InChI

1S/C6H11NS2/c1-4(2)5-3-9-6(8)7-5/h4-5H,3H2,1-2H3,(H,7,8)/t5-/m0/s1

InChI key

CWIZUGZKLJDJLE-YFKPBYRVSA-N

Application

A highly selective and efficient chiral auxiliary which can be directly reduced to its corresponding aldehyde and the chiral auxiliary by reductive cleavage with diisobutylaluminum hydride.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service