Documents

06513

Sigma-Aldrich

tert-Butyl formate

purum, ≥98.5% (GC)

Synonym(s):
Formic acid tert-butylester
Linear Formula:
HCOOC(CH3)3
CAS Number:
Molecular Weight:
102.13
Beilstein/REAXYS Number:
1738871
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum

assay

≥98.5% (GC)

refractive index

n20/D 1.379 (lit.)
n20/D 1.380

bp

82-83 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OC=O

InChI

1S/C5H10O2/c1-5(2,3)7-4-6/h4H,1-3H3

InChI key

RUPAXCPQAAOIPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Formylation of lactones

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Target Organs

Respiratory system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

F,Xi

Risk Statement

11-36/37

Safety Statement

9-16-24-33

RIDADR

UN 3272 3 / PGII

WGK Germany

WGK 3

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

W.R. Roush et al.
The Journal of Organic Chemistry, 45, 3927-3927 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.