MilliporeSigma
All Photos(1)

07187

Supelco

2,3-Dimethylpyrazine

analytical standard

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

refractive index

n20/D 1.505-1.509
n20/D 1.507 (lit.)

bp

156 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1nccnc1C

InChI

1S/C6H8N2/c1-5-6(2)8-4-3-7-5/h3-4H,1-2H3

InChI key

OXQOBQJCDNLAPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Protocols

GC Analysis of Volatiles in Roasted Coffee Beans on Omegawax® after SPME using a 50/30 µm DVB/Carboxen/PDMS Fiber

-Cymene; 2,5-Dimethylpyrrole; Acetoin, ≥96%, FCC, FG; 2,5-Dimethylpyrazine; 2,6-Dimethylpyrazine; 2-Ethylpyrazine, ≥98%, FG; 2,3-Dimethylpyrazine; 4-Heptanone; 3-Ethylpyridine; 2,3,5-Trimethylpyrazine; Furfural; Pyrrole; Furfuryl acetate; Linalool; Linalyl acetate; 5-Methylfurfural; γ-Butyrolactone; 2-Acetyl-1-methylpyrrole; Furfuryl alcohol; 2-Acetylpyrrole; Pyrrole-2-carboxaldehyde

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service