MilliporeSigma
All Photos(1)

07220

Sigma-Aldrich

DL-2-Aminobutyric acid

puriss., ≥99.0% (NT)

Linear Formula:
C2H5CH(NH2)CO2H
CAS Number:
Molecular Weight:
103.12
Beilstein:
635889
EC Number:
MDL number:
PubChem Substance ID:

grade

puriss.

assay

≥99.0% (NT)

mp

291 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CCC(N)C(O)=O

InChI

1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)

InChI key

QWCKQJZIFLGMSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service