110191

Sigma-Aldrich

Sodium acetate

ReagentPlus®, 99%

Synonym(s):
Acetic acid sodium salt
Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein/REAXYS Number:
3595639
EC Number:
MDL number:
eCl@ss:
39021906
PubChem Substance ID:
Pricing and availability is not currently available.

product line

ReagentPlus®

assay

99%

form

solid

autoignition temp.

1112 °F

pH

8.5-9.9 (25 °C, 246 g/L)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

solubility

ethanol: soluble(lit.)
water: soluble(lit.)

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

1

Flash Point(F)

>482 °F

Flash Point(C)

>250 °C

Certificate of Analysis
Certificate of Origin
PREPARATION OF (S)-2-Allyl-2-methylcyclohexanone (Cyclohexanone, 2-methyl-2-(2-propen-1-yl)-, (2S)-).
Justin T Mohr et al.
Organic syntheses; an annual publication of satisfactory methods for the preparation of organic chemicals, 86, 194-211 (2009-01-01)
The liquid-chromatographic quantification of some synthetic colorants in soft drinks.
Trandafir I, et al.
Scientific Study and Research, 10(1), 73-82 (2009)
Listeria monocytogenes inhibition in refrigerated vacuum packaged turkey bologna by chemical additives.
Wederquist HJ, et al.
Journal of Food Science, 59(3), 498-500 (1994)
R E Kingston et al.
Current protocols in molecular biology, Chapter 4, Unit4-Unit4 (2008-02-12)
Three different methods for RNA preparation using guanidine are presented in this unit--a single-step isolation method employing liquid-phase separation to selectively extract total RNA from tissues and cultured cells and two methods that rely on a CsCl step gradient to...
Eagleson M.
Concise Encyclopedia Chemistry, 995-995 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.