All Photos(1)

11218

Sigma-Aldrich

Ammonium nitrate

technical, granulated, with addition of anti-caking agent, ≥98% (alkalimetric)

Linear Formula:
NH4NO3
CAS Number:
Molecular Weight:
80.04
EC Number:
MDL number:
PubChem Substance ID:

grade

technical

assay

≥98% (alkalimetric)

quality

granulated
with addition of anti-caking agent

impurities

≤0.005% heavy metals (as Pb)

ign. residue

≤1% (as SO4)

bp

210 °C (lit.)

mp

169 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Fe: ≤20 mg/kg

SMILES string

N.O[N+]([O-])=O

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3

InChI key

PRORZGWHZXZQMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service