MilliporeSigma
All Photos(1)

12635

Sigma-Aldrich

Potassium chlorate

purified, ≥98.5%, powder

Linear Formula:
KClO3
CAS Number:
Molecular Weight:
122.55
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

assay

≥98.5%

form

powder

quality

purified

impurities

≤0.005% heavy metals (as Pb)

mp

356 °C (lit.)

anion traces

chloride (Cl-): ≤500 mg/kg

cation traces

Fe: ≤50 mg/kg

SMILES string

[K+].[O-]Cl(=O)=O

InChI

1S/ClHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

VKJKEPKFPUWCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service