All Photos(1)
26729
7000 series repair kits
for use with Hamilton 7000.5 KH
material
blunt tip stainless steel needle (point style 3)
needle size
25 ga (blunt tip)
description
7000.5KH
feature
(Kit includes needle, plunger wire, ferrule, and instructions.)
packaging
pkg of 1 ea
needle L
70 mm (2.75 in.)
volume
5 μL
compatibility
for use with Hamilton 7000.5 KH
General description
Kit includes needle, plunger wire, ferrule, and instructions.
Certificate of Analysis
Certificate of Origin