All Photos(1)

26732-U

7000 series repair kits

for use with Hamilton 7001 KH

NACRES:
NB.03

material

blunt tip stainless steel needle (point style 3)

needle size

25s ga (blunt tip)

description

7001KH

feature

(Kit includes needle, plunger wire, ferrule, and instructions.)

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 17188

needle L

70 mm (2.75 in.)

volume

1 μL

compatibility

for use with Hamilton 7001 KH

Looking for similar products? Visit Product Comparison Guide

General description

Kit includes needle, plunger wire, ferrule, and instructions.

Storage Class Code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service