MilliporeSigma
All Photos(1)

36030

Sigma-Aldrich

2,4-Dichloro-1-nitrobenzene

purum, ≥98.0% (GC)

Synonym(s):
1,3-Dichloro-4-nitrobenzene, asym.-Nitro-m-dichlorobenzene
Linear Formula:
Cl2C6H3NO2
CAS Number:
Molecular Weight:
192.00
Beilstein:
1451655
EC Number:
MDL number:
PubChem Substance ID:

grade

purum

assay

≥98.0% (GC)

bp

258 °C (lit.)

mp

29-32 °C (lit.)
30-32 °C

SMILES string

[O-][N+](=O)c1ccc(Cl)cc1Cl

InChI

1S/C6H3Cl2NO2/c7-4-1-2-6(9(10)11)5(8)3-4/h1-3H

InChI key

QUIMTLZDMCNYGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service