36607

Supelco

2,4-Dimethylphenol

PESTANAL®, analytical standard

Synonym(s):
asym.-m-Xylenol, 4-Hydroxy-m-xylene
Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Beilstein/REAXYS Number:
636244
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

vapor pressure

0.1 mmHg ( 25 °C)

product line

PESTANAL®

form

neat

shelf life

limited shelf life, expiry date on the label

application(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.538 (lit.)

bp

211-212 °C (lit.)

mp

22-23 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

Featured Industry

Agriculture
Cleaning Products
Cosmetics
Environmental
Flavors and Fragrances
Food and Beverages
Personal Care

format

neat

SMILES string

Cc1ccc(O)c(C)c1

InChI

1S/C8H10O/c1-6-3-4-8(9)7(2)5-6/h3-5,9H,1-2H3

InChI key

KUFFULVDNCHOFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Signal Word

Danger

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

T,N

Risk Statement

24/25-34-51/53

Safety Statement

26-36/37/39-45-61

RIDADR

UN 2261 6.1 / PGII

WGK Germany

3

Flash Point(F)

201.2 °F

Flash Point(C)

94 °C

S M Chernykh et al.
Prikladnaia biokhimiia i mikrobiologiia, 41(5), 578-581 (2005-10-26)
We optimized the conditions for production of laccase by lignolytic fungi Panus tigrinus 8/18. 2,4-Dimethylphenol was used as an aromatic inductor. The addition of 2,4-dimethylphenol and 2 mM CuSO4 to a rich medium was followed by a tenfold increase in...
Diversity-oriented synthesis of polycyclic scaffolds by modification of an anodic product derived from 2,4-dimethylphenol.
Joaquin Barjau et al.
Angewandte Chemie (International ed. in English), 50(6), 1415-1419 (2011-02-04)
F L Arenghi et al.
Applied and environmental microbiology, 67(7), 3304-3308 (2001-06-27)
Pseudomonas stutzeri OX1 meta pathway genes for toluene and o-xylene catabolism were analyzed, and loci encoding phenol hydroxylase, catechol 2,3-dioxygenase, 2-hydroxymuconate semialdehyde dehydrogenase, and 2-hydroxymuconate semialdehyde hydrolase were mapped. Phenol hydroxylase converted a broad range of substrates, as it was...
Hiroko Tsukatani et al.
Analytica chimica acta, 682(1-2), 72-76 (2010-11-09)
Gas chromatography/supersonic jet/resonance-enhanced multiphoton ionization/time-of-flight mass spectrometry (GC/SSJ/REMPI/TOF-MS) was employed for isomer-selective determination of 2,4-xylenol in river and seawater samples. The sample containing 2,4-xylenol was measured using argon, rather than helium, as the GC carrier gas to cool the analyte...
Peipei Qi et al.
Talanta, 81(4-5), 1630-1635 (2010-05-06)
A molecularly imprinted polymer (MIP) was prepared using 2,4-dimethylphenol (2,4-DMP) as template. The synthesis is optimized by using three different porogens, chloroform, acetonitrile and toluene. The MIP was used as a class-selective sorbent in molecularly imprinted solid-phase extraction (MIP-SPE) for...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.