Documents

41711

Supelco

2′,4′,6′-Trihydroxyacetophenone monohydrate

puriss. p.a., matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure

Synonym(s):
2-Acetylphloroglucinol, THAP
Linear Formula:
(HO)3C6H2COCH3·H2O
CAS Number:
Molecular Weight:
186.16
Beilstein/REAXYS Number:
9288656
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

matrix substance for MALDI-MS
puriss. p.a.

assay

≥99.5% (HPLC)

mp

219-221 °C (lit.)

cation traces

Ca: ≤200 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

in accordance for UV test

SMILES string

O.CC(=O)c1c(O)cc(O)cc1O

InChI

1S/C8H8O4.H2O/c1-4(9)8-6(11)2-5(10)3-7(8)12;/h2-3,10-12H,1H3;1H2

InChI key

GDSIBPPJKSBCMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.