51684

Millipore

Potato Dextrose Agar

suitable for microbiology, ready to use media in the bottle

Synonym(s):
Potato Glucose Agar
Pricing and availability is not currently available.

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
dextrose, 20 g/L
potato extract, 4 g/L

application(s)

microbiology: suitable

final pH

5.6±0.2 (25 °C)

Featured Industry

Agriculture
Environmental
Food and Beverages

storage temp.

2-8°C

suitability

nonselective for Aspergillus spp.
nonselective for Candida spp.
nonselective for Penicillium spp.
nonselective for Pichia spp.
nonselective for Saccharomyces spp.
nonselective for Zygosaccharomyces spp.
nonselective for molds (General Media)
nonselective for yeasts (General Media)

Looking for similar products? Visit Product Comparison Guide

Application

Recommended for the isolation and enumeration of yeasts and molds in dairy products and food.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

2

Certificate of Analysis
Certificate of Origin
Julia Schumacher et al.
Eukaryotic cell, 7(4), 584-601 (2008-02-12)
Recently, we showed that the alpha subunit BCG1 of a heterotrimeric G protein is an upstream activator of the Ca(2+)/calmodulin-dependent phosphatase calcineurin in the gray mold fungus Botrytis cinerea. To identify the transcription factor acting downstream of BCG1 and calcineurin...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.