All Photos(1)

546135

Sigma-Aldrich

Coumarin 6H

Dye content 99 %

Synonym(s):
2,3,5,6-1H,4H-Tetrahydroquinolizino[9,9a,1-gh]coumarin
Empirical Formula (Hill Notation):
C15H15NO2
CAS Number:
Molecular Weight:
241.29
EC Number:
MDL number:
PubChem Substance ID:

composition

Dye content, 99%

Quality Level

mp

130-132 °C (lit.)

λmax

394 nm

fluorescence

λem 476 nm in ethanol (Lasing peak 491nm, lasing range 463 - 522 nm (ethanol), pump source XeCl (308nm))

SMILES string

O=C1Oc2c3CCCN4CCCc(cc2C=C1)c34

InChI

1S/C15H15NO2/c17-13-6-5-11-9-10-3-1-7-16-8-2-4-12(14(10)16)15(11)18-13/h5-6,9H,1-4,7-8H2

InChI key

MZSOXGPKUOAXNY-UHFFFAOYSA-N

Packaging

100 mg in glass insert

Features and Benefits

Laser dye

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service