55817

Sigma-Aldrich

Fluorescent red 630

≥85% (HPLC)

Empirical Formula (Hill Notation):
C36H46N2O6S
CAS Number:
Molecular Weight:
634.83
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

assay

≥85% (HPLC)

fluorescence

λex 620 nm; λem 650 nm in 0.1 M phosphate pH 7.0

storage temp.

2-8°C

SMILES string

CCN(CC)c1ccc2c(OC(=C\C2=C\C=C\C3=N+(CCCCCC(O)=O)c4ccc(cc4C3(C)C)S(O-)(=O)=O)C(C)(C)C)c1

Other Notes

Stable near-IR fluorescent marker dye

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.