60980

Sigma-Aldrich

Suberonitrile

purum, ≥98.0% (GC)

Synonym(s):
Hexamethylene dicyanide, Octanedinitrile
Linear Formula:
NC(CH2)6CN
CAS Number:
Molecular Weight:
136.19
Beilstein/REAXYS Number:
1744365
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum

assay

≥98.0% (GC)

refractive index

n20/D 1.444 (lit.)

bp

185 °C/15 mmHg (lit.)

mp

−3.5 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

N#CCCCCCCC#N

InChI

1S/C8H12N2/c9-7-5-3-1-2-4-6-8-10/h1-6H2

InChI key

BTNXBLUGMAMSSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

T

Risk Statement

23/24/25

Safety Statement

36/37/39-45

RIDADR

UN 3276 6.1 / PGIII

WGK Germany

1

Flash Point(F)

230 °F

Flash Point(C)

110 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.