62558

Supelco

Lithium ionophore III

Selectophore, function tested

Synonym(s):
N,N-Dicyclohexyl-N′,N′-diisobutyl-cis-cyclohexane-1,2-dicarboxamide, ETH 1810
Empirical Formula (Hill Notation):
C28H50N2O2
CAS Number:
Molecular Weight:
446.71
Beilstein/REAXYS Number:
6536266
MDL number:
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

quality

function tested

mp

110-112 °C

SMILES string

CC(C)CN(CC(C)C)C(=O)[C@H]1CCCC[C@H]1C(=O)N(C2CCCCC2)C3CCCCC3

InChI

1S/C28H50N2O2/c1-21(2)19-29(20-22(3)4)27(31)25-17-11-12-18-26(25)28(32)30(23-13-7-5-8-14-23)24-15-9-6-10-16-24/h21-26H,5-20H2,1-4H3/t25-,26+/m0/s1

InChI key

QGKITQHJONXIQV-IZZNHLLZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Lithium ionophore used in liquid membrane electrodes for the determination of Li concentration in blood

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

A Zeevi et al.
The Journal of membrane biology, 121(2), 133-140 (1991-04-01)
The neutral, noncyclic Li(+)-selective ionophore ETH1810, which is a di-imide, differs structurally from previous similar ionophores by removal of the intramolecular symmetry of the N-imide substituents. Properties of this ionophore, as a potential carrier of lithium, were probed through studies...
Lithium/sodium ion concentration ratio measurements in blood serum with lithium and sodium ion selective liquid membrane electrodes.
E Metzger et al.
Analytical chemistry, 59(13), 1600-1603 (1987-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.