70450

Sigma-Aldrich

2-Naphthol

fluorescence indicator, ≥99.0%

Synonym(s):
β-Naphthol, 2-Hydroxynaphthalene
Linear Formula:
C10H7OH
CAS Number:
Molecular Weight:
144.17
Beilstein/REAXYS Number:
742134
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
MA.02
Pricing and availability is not currently available.

Quality Level

grade

fluorescence indicator

vapor density

4.97 (vs air)

vapor pressure

10 mmHg ( 145.5 °C)

assay

≥99.0% (GC)
≥99.0%

impurities

≤0.5% 1-naphthol

ign. residue

≤0.05%

bp

285-286 °C (lit.)

mp

120-122 °C (lit.)
121-122 °C

fluorescence

λex 320 nm; λem 410 nm in 0.1 M NaOH

SMILES string

Oc1ccc2ccccc2c1

InChI

1S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H

InChI key

JWAZRIHNYRIHIV-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

2-Naphthol (2OH) is a hydroxyarene molecule, which when electronically excited forms strong acid. Excited 2OH dissociates only in water. It has a slight phenolic odor. It is incompatible with strong oxidizing agents, strong bases, acid chlorides and acid anhydrides. It is one of the most commonly used fluorescence dye.

Application

2OH has been used as a fluorescence indicator which changes fluorescence intensity at varied temperatures when embedded in aqueous Poly(vinly alcohol)/borax gel networks.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3077 9 / PGIII

WGK Germany

WGK 2

Flash Point(F)

307.4 °F - closed cup

Flash Point(C)

153 °C - closed cup

Armour, Margaret-Ann
Hazardous Laboratory Chemicals Disposal Guide, 325-325 (2003)
Solvatochromism of ?-Naphthol
Solntsev, Kyril M., Dan Huppert, and Noam Agmon
The Journal of Physical Chemistry A, 102.47, 9599-9606 (1998)
A novel fluorescence temperature sensor based on a surfactant-free PVA/borax/2-naphthol hydrogel network system.
Lee, Sang Min, et al.
Journal of Applied Polymer Science, 93.5, 2114-2118 (2004)
Shouning Chai et al.
Environmental science & technology, 46(18), 10182-10190 (2012-08-28)
With in situ molecular imprinting technique, a novel nanoelectrode (MI, n-P)-TiO(2) with n-P heterojunction and molecular recognition ability was fabricated by liquid phase deposition at low temperature. Using bisphenol A (BPA) as template, the spindle-like TiO(2) particles 40-80 nm in...
Takuya Oguma et al.
Journal of the American Chemical Society, 134(49), 20017-20020 (2012-11-28)
Iron(salan) complex 1 was found to catalyze the oxidative dearomatization of 1-substituted 2-naphthols with the formation of an all-carbon quaternary stereocenter in air in the presence of nitroalkanes, to afford the corresponding cyclic enones with high enantioselectivity of 88-96% ee.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.