76950

Sigma-Aldrich

3-Pentanol

purum, ≥98.0% (GC)

Synonym(s):
sec-inact.-Pentyl alcohol, Diethyl carbinol
Linear Formula:
(C2H5)2CHOH
CAS Number:
Molecular Weight:
88.15
Beilstein/REAXYS Number:
1730964
EC Number:
MDL number:
PubChem Substance ID:

grade

purum

assay

≥98.0% (GC)

autoignition temp.

815 °F

refractive index

n20/D 1.410 (lit.)
n20/D 1.410

bp

114-115 °C/749 mmHg (lit.)

density

0.815 g/mL at 25 °C (lit.)

SMILES string

CCC(O)CC

InChI

1S/C5H12O/c1-3-5(6)4-2/h5-6H,3-4H2,1-2H3

InChI key

AQIXEPGDORPWBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service