All Photos(1)

78466

Supelco

Benzyl propionate

analytical standard

Linear Formula:
C2H5CO2CH2C6H5
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

grade

analytical standard

assay

≥98.5% (GC)

form

neat

shelf life

limited shelf life, expiry date on the label

application(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.496-1.50
n20/D 1.497 (lit.)

bp

222 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

Featured Industry

Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care

format

neat

SMILES string

CCC(=O)OCc1ccccc1

InChI

1S/C10H12O2/c1-2-10(11)12-8-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

VHOMAPWVLKRQAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service